Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Potassium tripolyphosphate 96%

Supplier: AMBEED, INC
Warning

A209165-1000G A209165-100G A209165-500G
76721-652EA 42 USD InStock
76721-652 76721-654 76721-656
Potassium tripolyphosphate 96%
Potassium tripolyphosphate
Precaución: for Research Use Only


Formula: K₅O₁₀P₃
MDL Number: MFCD00054129
CAS Number: 13845-36-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty