Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Trifluoroacetyl-3-methyl-1-phenyl-5-pyrazolone 98%

Supplier: AMBEED, INC
Warning

A219520-1G A219520-250MG A219520-5G
76721-932EA 63 USD InStock
76721-932 76721-934 76721-936
4-Trifluoroacetyl-3-methyl-1-phenyl-5-pyrazolone 98%
4-Trifluoroacetyl-3-methyl-1-phenyl-5-pyrazolone
Precaución: for Research Use Only


Formula: C₁₂H₉F₃N₂O₂
MDL Number: MFCD00143378
CAS Number: 1691-93-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty