Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(Trimethylsilyl)imidazole 98%

Supplier: AMBEED, INC
Danger

A292420-25G A292420-5G A292420-100G A292420-10G
76723-508EA 14 USD InStock
76723-508 76723-510 77601-300 77668-294
1-(Trimethylsilyl)imidazole 98%
1-(Trimethylsilyl)imidazole
Precaución: for Research Use Only


Formula: C₆H₁₂N₂Si
MDL Number: MFCD00005280
CAS Number: 18156-74-6
UN: 1993
ADR: 3,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty