Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

SKF-96365 hydrochloride 98%

Supplier: AMBEED, INC
Warning

A380689-10MG A380689-1MG A380689-5MG
76724-868EA 83 USD InStock
76724-868 76724-870 76724-872
SKF-96365 hydrochloride 98%
SKF-96365 hydrochloride
Precaución: for Research Use Only


Formula: C₂₂H₂₆N₂O₃·HCl
MDL Number: MFCD00236407
CAS Number: 130495-35-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty