Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Methyl-D-glucamine hydrochloride 97%

Supplier: AMBEED, INC
Warning

A755677-1G A755677-25G A755677-5G
76728-168EA 13 USD InStock
76728-168 76728-170 76728-172
N-Methyl-D-glucamine hydrochloride 97%
N-Methyl-D-glucamine hydrochloride
Precaución: for Research Use Only


Formula: C₇H₁₇NO₅
MDL Number: MFCD00060164
CAS Number: 35564-86-4
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty