Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sarafloxacin 98%

Supplier: AMBEED, INC
Warning

A798069-100MG A798069-250MG A798069-1G
76728-528EA 61 USD InStock
76728-528 76728-530 77670-122
Sarafloxacin 98%
Sarafloxacin
Precaución: for Research Use Only


Formula: C₂₀H₁₇F₂N₃O₃
MDL Number: MFCD00865974
CAS Number: 98105-99-8
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty