Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Methyl-1H-indazole-3-carbaldehyde 98%

Supplier: AMBEED, INC
Warning

A112553-100MG A112553-10G A112553-1G A112553-250MG
76765-958EA 14 USD InStock
76765-958 76765-960 76765-962 76765-964
5-Methyl-1H-indazole-3-carbaldehyde 98%
5-Methyl-1H-indazole-3-carbaldehyde
Precaución: for Research Use Only


Formula: C₉H₈N₂O
MDL Number: MFCD07781624
CAS Number: 518987-35-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty