Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(4-Chlorophenyl)cyclobutanecarbonitrile 98%

Supplier: AMBEED, INC
Warning

A129395-10G A129395-1G A129395-250MG A129395-25G A129395-5G
76767-108EA 47 USD InStock
76767-108 76767-110 76767-112 76767-114 76767-116
1-(4-Chlorophenyl)cyclobutanecarbonitrile 98%
1-(4-Chlorophenyl)cyclobutanecarbonitrile
Precaución: for Research Use Only


Formula: C₁₁H₁₀ClN
MDL Number: MFCD00065239
CAS Number: 28049-61-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty