Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Potassium-5-methyl-1,3,4-oxadiazole-2-carboxylate 95%

Supplier: AMBEED, INC
Warning

A138348-1G A138348-25G A138348-5G
76767-646EA 5 USD InStock
76767-646 76767-648 76767-650
Potassium-5-methyl-1,3,4-oxadiazole-2-carboxylate 95%
Potassium-5-methyl-1,3,4-oxadiazole-2-carboxylate
Precaución: for Research Use Only


Formula: C4H3KN2O3
MDL Number: MFCD08060085
CAS Number: 888504-28-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty