Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chloro-2-methylphenylboronic acid 98%

Supplier: AMBEED, INC
Warning

A153861-10G A153861-1G A153861-25G A153861-5G A153861-100G
76768-252EA 35 USD InStock
76768-252 76768-254 76768-256 76768-258 77625-338
3-Chloro-2-methylphenylboronic acid 98%
3-Chloro-2-methylphenylboronic acid
Precaución: for Research Use Only


Formula: C₇H₈BClO₂
MDL Number: MFCD04115641
CAS Number: 313545-20-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty