Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4'-Chloro-2,2,2-trifluoroacetophenone 97%

Supplier: AMBEED, INC
Warning

A163043-1G A163043-250MG A163043-25G
76768-664EA 16 USD InStock
76768-664 76768-666 77625-590
4'-Chloro-2,2,2-trifluoroacetophenone 97%
4'-Chloro-2,2,2-trifluoroacetophenone
Precaución: for Research Use Only


Formula: C₈H₄ClF₃O
MDL Number: MFCD00018686
CAS Number: 321-37-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty