Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Bicyclo[4.2.0]octa-1,3,5-trien-3-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 95%

Supplier: AMBEED, INC
Warning

A1682132-100MG A1682132-1G A1682132-250MG A1682132-5G
76769-052EA 48 USD InStock
76769-052 76769-054 76769-056 76769-058
2-(Bicyclo[4.2.0]octa-1,3,5-trien-3-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 95%
2-(Bicyclo[4.2.0]octa-1,3,5-trien-3-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Precaución: for Research Use Only


Formula: C₁₄H₁₉BO₂
CAS Number: 866756-09-4
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty