3,4-Dihydroxybenzonitrile 97%

Supplier: AMBEED, INC
Warning

A192395-100G A192395-25G A192395-500G A192395-5G
76769-912EA 123 USD
76769-912 76769-914 76769-916 76769-918 76769-920
3,4-Dihydroxybenzonitrile 97%
3,4-Dihydroxybenzonitrile

Precaución: for Research Use Only

Formula: C₇H₅NO₂
MW: 135.12
Boiling Pt: 311
MDL Number: MFCD00016436
CAS Number: 17345-61-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR