Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Phenyl-1,3-thiazol-2-amine 98%

Supplier: AMBEED, INC
Warning

A202180-10G A202180-1G A202180-250MG A202180-25G A202180-5G
76770-186EA 163 USD InStock
76770-186 76770-188 76770-190 76770-192 76770-194
5-Phenyl-1,3-thiazol-2-amine 98%
5-Phenyl-1,3-thiazol-2-amine
Precaución: for Research Use Only


Formula: C₉H₈N₂S
MDL Number: MFCD00022451
CAS Number: 39136-63-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty