Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Trifluoromethyl)-2-pyridinecarbonitrile 98%

Supplier: AMBEED, INC
Warning

A286604-100MG A286604-1G A286604-250MG
76772-406EA 15 USD InStock
76772-406 77626-474 77626-476
4-(Trifluoromethyl)-2-pyridinecarbonitrile 98%
4-(Trifluoromethyl)-2-pyridinecarbonitrile
Precaución: for Research Use Only


Formula: C₇H₃F₃N₂
MDL Number: MFCD09038236
CAS Number: 936841-69-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty