Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(5-Bromo-2-pyridinyl)-4-methylpiperazine 98%

Supplier: AMBEED, INC
Warning

A324514-250MG A324514-5G A324514-1G
76772-974EA 11 USD InStock
76772-974 76772-976 77626-604
1-(5-Bromo-2-pyridinyl)-4-methylpiperazine 98%
1-(5-Bromo-2-pyridinyl)-4-methylpiperazine
Precaución: for Research Use Only


Formula: C₁₀H₁₄BrN₃
MDL Number: MFCD07368225
CAS Number: 364794-58-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty