Ethyl-4-amino-1-piperidinecarboxylate 95%

Supplier: AMBEED, INC
Warning

A634815-100G A634815-25G A634815-5G
76775-478EA 205 USD
76775-478 76775-480 76775-482
Ethyl-4-amino-1-piperidinecarboxylate 95%
Ethyl-4-amino-1-piperidinecarboxylate

Precaución: for Research Use Only

Formula: C₈H₁₆N₂O₂
MW: 172.23 g/mol
MDL Number: MFCD00006484
CAS Number: 58859-46-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR