Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Methoxy-N-methyl-2-nitroaniline 95%

Supplier: AMBEED, INC
Warning

A657814-1G A657814-5G
76775-606EA 25 USD InStock
76775-606 76775-608
5-Methoxy-N-methyl-2-nitroaniline 95%
5-Methoxy-N-methyl-2-nitroaniline
Precaución: for Research Use Only


Formula: C₈H₁₀N₂O₃
MDL Number: MFCD05865077
CAS Number: 69397-93-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty