Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diallyl propyl isocyanurate 95% stabilized

Supplier: AMBEED, INC
Warning

Stabilization: Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)

A659187-1G A659187-250MG
76775-616EA 15 USD InStock
76775-616 76775-618
Diallyl propyl isocyanurate 95% stabilized
Diallyl propyl isocyanurate
Precaución: for Research Use Only


Formula: C₁₂H₁₇N₃O₃
MDL Number: MFCD00144514
CAS Number: 5320-25-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty