Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Chloroimidazo[1,2-b]pyridazine hydrochloride 98%

Supplier: AMBEED, INC
Warning

A787955-100G A787955-25G A787955-5G
76776-486EA 40 USD InStock
76776-486 76776-488 76776-490
6-Chloroimidazo[1,2-b]pyridazine hydrochloride 98%
6-Chloroimidazo[1,2-b]pyridazine hydrochloride
Precaución: for Research Use Only


Formula: C₆H₅Cl₂N₃
MDL Number: MFCD09258637
CAS Number: 13493-79-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty