Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2R,3S,4R,5R)-2-(Hydroxymethyl)-5-(6-mercapto-9H-purin-9-yl)tetrahydrofuran-3,4-diol 95%

Supplier: AMBEED, INC
Warning

A792872-100MG A792872-250MG
76776-498EA 29 USD InStock
76776-498 76776-500
(2R,3S,4R,5R)-2-(Hydroxymethyl)-5-(6-mercapto-9H-purin-9-yl)tetrahydrofuran-3,4-diol 95%
(2R,3S,4R,5R)-2-(Hydroxymethyl)-5-(6-mercapto-9H-purin-9-yl)tetrahydrofuran-3,4-diol
Precaución: for Research Use Only


Formula: C₁₀H₁₂N₄O₄S
MDL Number: MFCD00005743
CAS Number: 574-25-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty