Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dibenzo-21-crown-7-(dibenzo-1,4,7,10,13,16,19-heptaoxacycloheneicosane) 96%

Supplier: AMBEED, INC
Warning

A816441-100MG A816441-1G A816441-250MG
76776-584EA 18 USD InStock
76776-584 76776-586 76776-588
Dibenzo-21-crown-7-(dibenzo-1,4,7,10,13,16,19-heptaoxacycloheneicosane) 96%
Dibenzo-21-crown-7-(dibenzo-1,4,7,10,13,16,19-heptaoxacycloheneicosane)
Precaución: for Research Use Only


Formula: C₂₂H₂₈O₇
MDL Number: MFCD00068646
CAS Number: 14098-41-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty