Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-(Dibutylamino)-1,3,5-triazine-2,4-dithiol 98%

Supplier: AMBEED, INC
Danger

A995717-100G A995717-25G A995717-5G
76777-082EA 83 USD InStock
76777-082 76777-084 76777-086
6-(Dibutylamino)-1,3,5-triazine-2,4-dithiol 98%
6-(Dibutylamino)-1,3,5-triazine-2,4-dithiol
Precaución: for Research Use Only


Formula: C₁₁H₂₀N₄S₂
MDL Number: MFCD00272594
CAS Number: 29529-99-5
UN: 3077
ADR: 9,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty