Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

H-DAB(boc)-OMe·HCl 97%

Supplier: AMBEED, INC
Warning

A104176-10G A104176-250MG A104176-25G A104176-5G
76780-676EA 137 USD InStock
76780-676 76780-678 76780-680 76780-682
H-DAB(boc)-OMe·HCl 97%
H-DAB(boc)-OMe·HCl
Precaución: for Research Use Only


Formula: C₁₀H₂₁ClN₂O₄
MDL Number: MFCD00798680
CAS Number: 3350-15-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty