Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Amino-2,4,6-trichloropyrimidine 98%

Supplier: AMBEED, INC
Warning

A130126-100MG A130126-250MG
76782-310EA 39 USD InStock
76782-310 76782-312
5-Amino-2,4,6-trichloropyrimidine 98%
5-Amino-2,4,6-trichloropyrimidine
Precaución: for Research Use Only


Formula: C₄H₂Cl₃N₃
MDL Number: MFCD08693695
CAS Number: 91322-00-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty