4-(4-Chlorophenoxy)phenol 98%

Supplier: AMBEED, INC
Warning

A138451-25G A138451-5G
76782-798EA 102 USD
76782-798 76782-800
4-(4-Chlorophenoxy)phenol 98%
4-(4-Chlorophenoxy)phenol

Precaución: for Research Use Only

Formula: C₁₂H₉ClO₂
MW: 220.65 g/mol
MDL Number: MFCD00125839
CAS Number: 21567-18-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR