4-(2-Hydroxyphenyl)phenol 98%

Supplier: AMBEED, INC
Danger

A160568-1G A160568-250MG
76783-810EA 292 USD
76783-810 76783-812
4-(2-Hydroxyphenyl)phenol 98%
4-(2-Hydroxyphenyl)phenol

Precaución: for Research Use Only

Formula: C₁₂H₁₀O₂
MW: 186.207 g/mol
MDL Number: MFCD11895228
CAS Number: 611-62-1
UN: 3077
ADR: 9 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR