1-Phenyloxindole 98%

Supplier: AMBEED, INC
Warning

A162902-100G A162902-1G A162902-25G A162902-5G
76783-942EA 80 USD
76783-942 76783-944 76783-946 76783-948
1-Phenyloxindole 98%
1-Phenyloxindole

Precaución: for Research Use Only

Formula: C₁₄H₁₁NO
MW: 209.25 g/mol
MDL Number: MFCD00234850
CAS Number: 3335-98-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR