Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3'-Fluoro-2'-hydroxyacetophenone 97%

Supplier: AMBEED, INC
Warning

A261976-100MG A261976-1G A261976-250MG A261976-25G
76787-016EA 15 USD InStock
76787-016 76787-018 76787-020 76787-022
3'-Fluoro-2'-hydroxyacetophenone 97%
3'-Fluoro-2'-hydroxyacetophenone
Precaución: for Research Use Only


Formula: C₈H₇FO₂
MDL Number: MFCD09055140
CAS Number: 699-92-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty