Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-2-(4-fluorophenyl)acetate 97%

Supplier: AMBEED, INC
Warning

A508392-10G A508392-1G A508392-25G A508392-5G A508392-100G
76789-990EA 14 USD InStock
76789-990 76789-992 76789-994 76789-996 77669-272
Ethyl-2-(4-fluorophenyl)acetate 97%
Ethyl-2-(4-fluorophenyl)acetate
Precaución: for Research Use Only


Formula: C₁₀H₁₁FO₂
MW: 182.1916 g/mol
MDL Number: MFCD08235152
CAS Number: 587-88-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty