2-Acetamidofluorene 99+%

Supplier: AMBEED, INC
Warning

A534089-1G A534089-250MG A534089-25G A534089-5G
76790-174EA 47 USD
76790-174 76790-176 76790-178 76790-180
2-Acetamidofluorene 99+%
2-Acetamidofluorene

Precaución: for Research Use Only

Formula: C₁₅H₁₃NO
MW: 223.28 g/mol
MDL Number: MFCD00001116
CAS Number: 53-96-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR