Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tetraethylammonium hexafluorophosphate 98%

Supplier: AMBEED, INC
Warning

A590789-100G A590789-25G A590789-500G A590789-5G
76790-482EA 70 USD InStock
76790-482 76790-484 76790-486 76790-488
Tetraethylammonium hexafluorophosphate 98%
Tetraethylammonium hexafluorophosphate
Precaución: for Research Use Only


Formula: C₈H₂₀F₆NP
MDL Number: MFCD00043179
CAS Number: 429-07-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty