Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Pyrazolo[1,5-a]pyrimidin-7(4H)-one 98%

Supplier: AMBEED, INC
Warning

A619566-1G A619566-250MG A619566-25G A619566-5G
76790-652EA 53 USD InStock
76790-652 76790-654 76790-656 76790-658
Pyrazolo[1,5-a]pyrimidin-7(4H)-one 98%
Pyrazolo[1,5-a]pyrimidin-7(4H)-one
Precaución: for Research Use Only


Formula: C₆H₅N₃O
MDL Number: MFCD09746272
CAS Number: 29274-23-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty