Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

trans-4-(Hydroxymethyl)cyclohexanecarboxylic acid ≥97%

Supplier: AMBEED, INC
Warning

A846332-1G A846332-250MG A846332-5G
76791-868EA 55 USD InStock
76791-868 76791-870 77670-210
trans-4-(Hydroxymethyl)cyclohexanecarboxylic acid ≥97%
trans-4-(Hydroxymethyl)cyclohexanecarboxylic acid
Precaución: for Research Use Only


Formula: C₈H₁₄O₃
MDL Number: MFCD03840884
CAS Number: 66185-74-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty