Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 2,5-dibromopentanoate 95%

Supplier: AMBEED, INC
Danger

A106506-10G A106506-1G A106506-250MG A106506-5G
76796-558EA 96 USD InStock
76796-558 76796-560 76796-562 76796-564
Methyl 2,5-dibromopentanoate 95%
Methyl 2,5-dibromopentanoate
Precaución: for Research Use Only


Formula: C₆H₁₀Br₂O₂
MDL Number: MFCD02684289
CAS Number: 50995-48-7
UN: 3265
ADR: 8,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty