Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,4-Methylenedioxycinnamic acid 95%

Supplier: AMBEED, INC
Warning

A109120-100G A109120-10G A109120-25G A109120-500G
76796-716EA 53 USD InStock
76796-716 76796-718 76796-720 76796-722
3,4-Methylenedioxycinnamic acid 95%
3,4-Methylenedioxycinnamic acid
Precaución: for Research Use Only


Formula: C₁₀H₈O₄
MDL Number: MFCD00005837
CAS Number: 2373-80-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty