Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Lubiprostone 98%

Supplier: AMBEED, INC
Warning

A126294-10MG A126294-1MG A126294-5MG
76797-670EA 67 USD InStock
76797-670 76797-672 76797-674
Lubiprostone 98%
Lubiprostone
Precaución: for Research Use Only


Formula: C₂₀H₃₂F₂O₅
MDL Number: MFCD20268389
CAS Number: 136790-76-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty