Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl pentafluorobenzoate 98%

Supplier: AMBEED, INC
Warning

A139911-100G A139911-1G A139911-25G A139911-5G
76798-184EA 141 USD InStock
76798-184 76798-186 76798-188 76798-190
Ethyl pentafluorobenzoate 98%
Ethyl pentafluorobenzoate
Precaución: for Research Use Only


Formula: C₉H₅F₅O₂
MDL Number: MFCD00039211
CAS Number: 4522-93-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty