Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Acetyl-3,5-diiodo-L-tyrosine 97%

Supplier: AMBEED, INC
Warning

A142617-10G A142617-25G
76798-374EA 21 USD InStock
76798-374 76798-376
N-Acetyl-3,5-diiodo-L-tyrosine 97%
N-Acetyl-3,5-diiodo-L-tyrosine
Precaución: for Research Use Only


Formula: C₁₁H₁₁I₂NO₄
MDL Number: MFCD00020004
CAS Number: 1027-28-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty