N-((4-Amino-2-methylpyrimidin-5-yl)methyl)-N-(5-hydroxy-3-(((tetrahydrofuran-2-yl)methyl)disulfanyl)pent-2-en-2-yl)formamide 98%
Supplier: AMBEED, INC
|
Warning
|
A175577-100MG
A175577-25G
A175577-5G
A175577-1G
76799-524EA
5
USD
76799-524
76799-526
76799-528
77667-446
N-((4-Amino-2-methylpyrimidin-5-yl)methyl)-N-(5-hydroxy-3-(((tetrahydrofuran-2-yl)methyl)disulfanyl)pent-2-en-2-yl)formamide 98%
N-((4-Amino-2-methylpyrimidin-5-yl)methyl)-N-(5-hydroxy-3-(((tetrahydrofuran-2-yl)methyl)disulfanyl)pent-2-en-2-yl)formamide
Precaución: for Research Use Only
Formula:
C₁₇H₂₆N₄O₃S₂ MW: 398.54 g/mol |
MDL Number:
MFCD00867383 CAS Number: 804-30-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...