Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Nα-(2,4-Dinitro-5-fluorophenyl)-L-valinamide 97%

Supplier: AMBEED, INC
Warning

A178118-100MG A178118-1G A178118-250MG
76799-596EA 11 USD InStock
76799-596 76799-598 76799-600
Nα-(2,4-Dinitro-5-fluorophenyl)-L-valinamide 97%
Nα-(2,4-Dinitro-5-fluorophenyl)-L-valinamide
Precaución: for Research Use Only


Formula: C₁₁H₁₃FN₄O₅
MDL Number: MFCD00155636
CAS Number: 132679-61-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty