Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Nitro-1-naphthylamine 97%

Supplier: AMBEED, INC
Warning

A259619-10G A259619-1G A259619-25G A259619-500G A259619-5G A259619-100G
76801-750EA 25 USD InStock
76801-750 76801-752 76801-754 76801-756 76801-758 77626-352
4-Nitro-1-naphthylamine 97%
4-Nitro-1-naphthylamine
Precaución: for Research Use Only


Formula: C₁₀H₈N₂O₂
MDL Number: MFCD00004026
CAS Number: 776-34-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty