Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethosuximide 98%

Supplier: AMBEED, INC
Warning

A320514-1MG A320514-250MG A320514-5G
76802-894EA 5 USD InStock
76802-894 76802-896 77668-428
Ethosuximide 98%
Ethosuximide
Precaución: for Research Use Only


Formula: C₇H₁₁NO₂
MDL Number: MFCD00072123
CAS Number: 77-67-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty