Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Ethyl-3-hydroxy-N,N-dimethylbenzenaminium chloride 98%

Supplier: AMBEED, INC
Warning

A329103-250MG A329103-100MG A329103-1G
76803-014EA 5.31 USD InStock
76803-014 76803-016 76803-018 77626-622 77626-624
N-Ethyl-3-hydroxy-N,N-dimethylbenzenaminium chloride 98%
N-Ethyl-3-hydroxy-N,N-dimethylbenzenaminium chloride
Precaución: for Research Use Only


Formula: C₁₀H₁₆ClNO
MDL Number: MFCD00055064
CAS Number: 116-38-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty