Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium 1-hydroxypropane-1,2,3-tricarboxylate 97%

Supplier: AMBEED, INC
Warning

A549503-1G A549503-250MG A549503-5G
76805-468EA 47 USD InStock
76805-468 76805-470 76805-472
Sodium 1-hydroxypropane-1,2,3-tricarboxylate 97%
Sodium 1-hydroxypropane-1,2,3-tricarboxylate
Precaución: for Research Use Only


Formula: C₆H₅Na₃O₇
MDL Number: MFCD00004241
CAS Number: 1637-73-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty