Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Zinc(II) dibenzyldithiocarbamate 98% (by titrimetric analysis)

Supplier: AMBEED, INC
Danger

A552494-25G A552494-100G
76805-514EA 5 USD InStock
76805-514 77627-138
Zinc(II) dibenzyldithiocarbamate 98% (by titrimetric analysis)
Zinc(II) dibenzyldithiocarbamate
Precaución: for Research Use Only


Formula: C₁₅H₁₅NS₂
MDL Number: MFCD00014470
CAS Number: 14726-36-4
UN: 3077
ADR: 9,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty