Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Felbamate 98%

Supplier: AMBEED, INC
Warning

A637947-100MG A637947-10MG A637947-1MG A637947-25MG A637947-50MG
76806-284EA 122 USD InStock
76806-284 76806-286 76806-288 76806-290 76806-292
Felbamate 98%
Felbamate
Precaución: for Research Use Only


Formula: C₁₁H₁₄N₂O₄
MDL Number: MFCD00865296
CAS Number: 25451-15-4
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty