Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(Triethoxysilyl)methyl methacrylate 97% (by GC) stabilized

Supplier: AMBEED, INC
Warning

Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

A962429-25G A962429-5G
76808-126EA 110 USD InStock
76808-126 76808-128
(Triethoxysilyl)methyl methacrylate 97% (by GC) stabilized
(Triethoxysilyl)methyl methacrylate
Precaución: for Research Use Only


Formula: C₁₁H₂₂O₅Si
MDL Number: MFCD00156470
CAS Number: 5577-72-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty