Ethyl-1-methylpyrazole-4-carboxylate 97%

Supplier: AMBEED, INC
Warning

A107585-100G A107585-10G A107585-1G A107585-250MG A107585-25G A107585-5G
76809-760EA 288 USD
76809-760 76809-762 76809-764 76809-766 76809-768 76809-770
Ethyl-1-methylpyrazole-4-carboxylate 97%
Ethyl-1-methylpyrazole-4-carboxylate

Precaución: for Research Use Only

Formula: C₇H₁₀N₂O₂
MW: 154.17 g/mol
MDL Number: MFCD12180234
CAS Number: 85290-80-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR