Tofacitinib (CP-690550) citrate 99%
Supplier: AMBEED, INC
|
Warning
|
A118834-10MG
A118834-1MG
A118834-50MG
A118834-5MG
A118834-1G
A118834-100MG
76810-352EA
10
USD
76810-352
76810-354
76810-356
76810-358
77610-982
77670-674
Tofacitinib (CP-690550) citrate 99%
Tofacitinib (CP-690550) citrate
Precaución: For research use only.
Formula:
C₁₆H₂₀N₆O·C₆H₈O₇ MW: 504.49 g/mol |
MDL Number:
MFCD11616529 CAS Number: 540737-29-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...